missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-m-phenylenediamine, 97%

Catalog No. AAL0648506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0648506 5 g
AAL0648514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0648506 Supplier Thermo Scientific Chemicals Supplier No. L0648506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33786-89-9 | C6H7ClN2 | 142.586 g/mol

5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 33786-89-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00014775
InChI Key VZNUCJOYPXKLTA-UHFFFAOYSA-N
Synonym 5-chloro-m-phenylenediamine, 3,5-diaminochlorobenzene, 5-chloro-1,3-benzenediamine, 1,3-benzenediamine, 5-chloro, 5-chloro-1,3-phenylenediamine, ccris 2886, 5-amino-3-chloroaniline, acmc-20am42, 2-13-00-00029 beilstein handbook reference, 5-chloro-1,3-diaminobenzene
PubChem CID 73335
IUPAC Name 5-chlorobenzene-1,3-diamine
SMILES C1=C(C=C(C=C1N)Cl)N

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
UN Number UN1673
Beilstein 3029854
Solubility Information Slightly soluble in water.
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-m-phenylenediamine
Hazard Category H302+H312+H332-H341
Hazard Statement GHS H Statement
H331-H341-H302-H312
Toxic if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES
RTECSNumber CZ1579500
TSCA Yes

RUO – Research Use Only