accessibility menu, dialog, popup

UserName

NH2NH2Cl

CAS RN 33786-89-9

IUPAC Name: 5-chlorobenzene-1,3-diamine
Synonyms: 5-chlorobenzene-1,3-diamine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: VZNUCJOYPXKLTA-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)