missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-8-hydroxy-7-iodoquinoline 97.0+%, TCI America™

Catalog No. p-7124268
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C018725G 25 g
C0187250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C018725G Supplier TCI America Supplier No. C018725G
Only null left

Chemical Identifiers

CAS 130-26-7
Molecular Formula C9H5ClINO
Molecular Weight (g/mol) 305.499
MDL Number MFCD00006787
InChI Key QCDFBFJGMNKBDO-UHFFFAOYSA-N
Synonym clioquinol, iodochlorhydroxyquin, chloroiodoquin, chinoform, chloroiodoquine, 5-chloro-8-hydroxy-7-iodoquinoline, iodochloroxyquinoline, cliquinol, vioform, chlorojodochin
PubChem CID 2788
ChEBI CHEBI:74460
IUPAC Name 5-chloro-7-iodoquinolin-8-ol
SMILES C1=CC2=C(C(=C(C=C2Cl)I)O)N=C1

Specifications

Quantity 25 g
Formula Weight 305.50
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-8-hydroxy-7-iodoquinoline
RTECSNumber VC5075000
TSCA Yes