missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-7-methylisatin, 97%
CAS: 14389-06-1 | C9H6ClNO2 | 195.602 g/mol
Supplier: Thermo Scientific Chemicals L1168203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Chloro-7-methylisatin | |
| ∽283°C (decomposition) | |
| MFCD00047218 | |
| 153061 | |
| LDFQLYHDZZPAGN-UHFFFAOYSA-N | |
| 5-chloro-7-methyl-1H-indole-2,3-dione | |
| 366710 | |
| 97% |
| 14389-06-1 | |
| C9H6ClNO2 | |
| 1 g | |
| 5-chloro-7-methylisatin, 5-chloro-7-methylindoline-2,3-dione, 5-chloro-7-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-chloro-7-methyl-1h-benzo d azolidine-2,3-dione, pubchem13601, acmc-1buvx, isatin-based compound, 56, 5-chloro-7-methyl isatin, 1h-indole-2,3-dione,5-chloro-7-methyl, 5-chloranyl-7-methyl-1h-indole-2,3-dione | |
| CC1=C2C(=CC(=C1)Cl)C(=O)C(=O)N2 | |
| 195.602 | |
| 195.61 |
Chemical Identifiers
| 14389-06-1 | |
| 195.602 | |
| LDFQLYHDZZPAGN-UHFFFAOYSA-N | |
| 366710 | |
| CC1=C2C(=CC(=C1)Cl)C(=O)C(=O)N2 |
| C9H6ClNO2 | |
| MFCD00047218 | |
| 5-chloro-7-methylisatin, 5-chloro-7-methylindoline-2,3-dione, 5-chloro-7-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-chloro-7-methyl-1h-benzo d azolidine-2,3-dione, pubchem13601, acmc-1buvx, isatin-based compound, 56, 5-chloro-7-methyl isatin, 1h-indole-2,3-dione,5-chloro-7-methyl, 5-chloranyl-7-methyl-1h-indole-2,3-dione | |
| 5-chloro-7-methyl-1H-indole-2,3-dione |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only