missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-7-methylisatin, 97%

Catalog No. p-7045710
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL11682MD 250 mg
AAL1168203 1 g
AAL1168206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL11682MD Supplier Thermo Scientific Chemicals Supplier No. L11682MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14389-06-1 | C9H6ClNO2 | 195.602 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14389-06-1
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD00047218
InChI Key LDFQLYHDZZPAGN-UHFFFAOYSA-N
Synonym 5-chloro-7-methylisatin, 5-chloro-7-methylindoline-2,3-dione, 5-chloro-7-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-chloro-7-methyl-1h-benzo d azolidine-2,3-dione, pubchem13601, acmc-1buvx, isatin-based compound, 56, 5-chloro-7-methyl isatin, 1h-indole-2,3-dione,5-chloro-7-methyl, 5-chloranyl-7-methyl-1h-indole-2,3-dione
PubChem CID 366710
IUPAC Name 5-chloro-7-methyl-1H-indole-2,3-dione
SMILES CC1=C2C(=CC(=C1)Cl)C(=O)C(=O)N2

Specifications

Melting Point ∽283°C (decomposition)
Quantity 250 mg
Beilstein 153061
Formula Weight 195.61
Percent Purity 97%
Chemical Name or Material 5-Chloro-7-methylisatin
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only