missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-nitrosalicylaldehyde 98.0+%, TCI America™

Catalog No. C22495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22495G 5 g
C22491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22495G Supplier TCI America Supplier No. C22495G

Chemical Identifiers

CAS 16634-90-5
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00462959
InChI Key YSDCZOBSHGDCCI-UHFFFAOYSA-N
Synonym 5-Chloro-2-hydroxy-3-nitrobenzaldehyde
PubChem CID 177763
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)C=O)Cl

Specifications

Melting Point 110°C
Color Yellow
Quantity 5 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-3-nitrosalicylaldehyde