accessibility menu, dialog, popup

UserName

OHOClNOO

CAS RN 16634-90-5

IUPAC Name: 5-chloro-2-hydroxy-3-nitrobenzaldehyde
Synonyms: 5-chloro-2-hydroxy-3-nitrobenzaldehyde
Molecular Weight (g/mol): 201.56
Molecular Formula: C7H4ClNO4
InChi Key: YSDCZOBSHGDCCI-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=C(Cl)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results