missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-thiophenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America C234325G
Specifications
| 5-Chloro-2-thiophenesulfonyl Chloride | |
| 66°C | |
| MFCD00051667 | |
| 3265 | |
| SORSTNOXGOXWAO-UHFFFAOYSA-N | |
| 5-chlorothiophene-2-sulfonyl chloride | |
| 2733925 | |
| ≥98.0% (GC,T) |
| 2766-74-7 | |
| C4H2Cl2O2S2 | |
| 25 g | |
| 5-chlorothiophene-2-sulfonylchloride, 5-chloro-2-thiophenesulfonyl chloride, 5-chlorothiophene-2-sulphonyl chloride, 5-chlorothiophenesulphonyl chloride, 5-chloro-2-thienylsulfonyl chloride, 5-chloro-thiophene-2-sulfonyl chloride, 5-chlorothiophenesulfonyl chloride, 5-chlorothiophen-2-sulfonyl chloride, 5-chlorothiophene-2-sulphonylchloride, 2-thiophenesulfonyl chloride, 5-chloro | |
| ClC1=CC=C(S1)S(Cl)(=O)=O | |
| 217.08 | |
| 217.08 | |
| Liquid |
Chemical Identifiers
| 2766-74-7 | |
| 217.08 | |
| SORSTNOXGOXWAO-UHFFFAOYSA-N | |
| 2733925 | |
| ClC1=CC=C(S1)S(Cl)(=O)=O |
| C4H2Cl2O2S2 | |
| MFCD00051667 | |
| 5-chlorothiophene-2-sulfonylchloride, 5-chloro-2-thiophenesulfonyl chloride, 5-chlorothiophene-2-sulphonyl chloride, 5-chlorothiophenesulphonyl chloride, 5-chloro-2-thienylsulfonyl chloride, 5-chloro-thiophene-2-sulfonyl chloride, 5-chlorothiophenesulfonyl chloride, 5-chlorothiophen-2-sulfonyl chloride, 5-chlorothiophene-2-sulphonylchloride, 2-thiophenesulfonyl chloride, 5-chloro | |
| 5-chlorothiophene-2-sulfonyl chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator