missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-thiophenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7125422
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C23435G 5 g
C234325G 25 g
2 options

Catalog No. C23435G

Supplier: TCI America C23435G

Only null left

Chemical Identifiers

CAS 2766-74-7
Molecular Formula C4H2Cl2O2S2
Molecular Weight (g/mol) 217.08
MDL Number MFCD00051667
InChI Key SORSTNOXGOXWAO-UHFFFAOYSA-N
Synonym 5-chlorothiophene-2-sulfonylchloride, 5-chloro-2-thiophenesulfonyl chloride, 5-chlorothiophene-2-sulphonyl chloride, 5-chlorothiophenesulphonyl chloride, 5-chloro-2-thienylsulfonyl chloride, 5-chloro-thiophene-2-sulfonyl chloride, 5-chlorothiophenesulfonyl chloride, 5-chlorothiophen-2-sulfonyl chloride, 5-chlorothiophene-2-sulphonylchloride, 2-thiophenesulfonyl chloride, 5-chloro
PubChem CID 2733925
IUPAC Name 5-chlorothiophene-2-sulfonyl chloride
SMILES ClC1=CC=C(S1)S(Cl)(=O)=O

Specifications

Boiling Point 66°C
Quantity 5 g
UN Number 3265
Formula Weight 217.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-thiophenesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator