missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-thiophenesulfonamide 98.0+%, TCI America™

Catalog No. p-7125464
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24411G 1 g
C24415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24411G Supplier TCI America Supplier No. C24411G
Only null left

Chemical Identifiers

CAS 53595-66-7
Molecular Formula C4H4ClNO2S2
Molecular Weight (g/mol) 197.65
MDL Number MFCD00052584
InChI Key RKLQLYBJAZBSEU-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenesulfonamide, 2-thiophenesulfonamide, 5-chloro, 2-chloro thiophene-5-sulfonamide, pubchem7381, 5-chlorothiophenesulfonamide, 4-18-00-06708 beilstein handbook reference, ksc490i0r, 2-chloro-5-sulfamoylthiophene, 5-chloro-2-thienylsulphonamide
PubChem CID 1241301
IUPAC Name 5-chlorothiophene-2-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 117°C
Quantity 1 g
Formula Weight 197.65
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-thiophenesulfonamide
RTECSNumber XN0296000
TSCA No