missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-phenyl-1H-imidazole-4-carboxaldehyde 97.0+%, TCI America™

Catalog No. p-7125784
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30181G 1 g
C30185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30181G Supplier TCI America Supplier No. C30181G
Only null left

Chemical Identifiers

CAS 60367-52-4
Molecular Formula C10H7ClN2O
Molecular Weight (g/mol) 206.629
MDL Number MFCD01935953
InChI Key CCXUATWKEDQRMP-UHFFFAOYSA-N
Synonym 5-chloro-2-phenyl-3h-imidazole-4-carbaldehyde, 4-chloro-2-phenyl-1h-imidazole-5-carbaldehyde, 5-chloro-2-phenyl-1h-imiadazole-4-carbaldehyde, 5-chloro-2-phenylimidazole-4-carboxaldehyde, 1h-imidazole-4-carboxaldehyde, 5-chloro-2-phenyl, pubchem18833, ksc496q2r, chlorophenylimidazolecarbaldehyde, 2-phenyl-4-chloro-5-formylimidazole, 4-chloro-5-formyl-2-phenylimidazole
PubChem CID 2763376
IUPAC Name 5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde
SMILES C1=CC=C(C=C1)C2=NC(=C(N2)Cl)C=O

Specifications

Melting Point 171°C
Quantity 1 g
Formula Weight 206.63
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-phenyl-1H-imidazole-4-carboxaldehyde