CAS RN 60367-52-4
IUPAC Name:
5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde
Synonyms:
5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde
Molecular Weight (g/mol):
206.63
Molecular Formula:
C10H7ClN2O
InChi Key:
CCXUATWKEDQRMP-UHFFFAOYSA-N
SMILES:
ClC1=C(C=O)N=C(N1)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Chloro-2-phenyl-1H-imidazole-4-carboxaldehyde 97.0+%, TCI America™
CAS: 60367-52-4 Molecular Formula: C10H7ClN2O Molecular Weight (g/mol): 206.629 MDL Number: MFCD01935953 InChI Key: CCXUATWKEDQRMP-UHFFFAOYSA-N Synonym: 5-chloro-2-phenyl-3h-imidazole-4-carbaldehyde,4-chloro-2-phenyl-1h-imidazole-5-carbaldehyde,5-chloro-2-phenyl-1h-imiadazole-4-carbaldehyde,5-chloro-2-phenylimidazole-4-carboxaldehyde,1h-imidazole-4-carboxaldehyde, 5-chloro-2-phenyl,pubchem18833,ksc496q2r,chlorophenylimidazolecarbaldehyde,2-phenyl-4-chloro-5-formylimidazole,4-chloro-5-formyl-2-phenylimidazole PubChem CID: 2763376 IUPAC Name: 5-chloro-2-phenyl-1H-imidazole-4-carbaldehyde SMILES: C1=CC=C(C=C1)C2=NC(=C(N2)Cl)C=O