missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. C022425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C022425G 25 g
C0224250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C022425G Supplier TCI America Supplier No. C022425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2516-95-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007290
InChI Key ZKUYSJHXBFFGPU-UHFFFAOYSA-N
Synonym 5-chloro-2-nitrobenzoic acid, benzoic acid, 5-chloro-2-nitro, 2-nitro-5-chlorobenzoic acid, 3-chloro-6-nitrobenzoic acid, 5-chloro-2-nitro-benzoic acid, pubchem4586, acmc-209ghs, rarechem al bo 1106, timtec-bb sbb009922, 2-nitro-5-chlorobenzoate
PubChem CID 17286
SMILES C1=CC(=C(C=C1Cl)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 139°C
Color Yellow
Quantity 25 g
Formula Weight 201.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitrobenzoic Acid
EINECSNumber (3)-1502
TSCA Yes