missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-nitrobenzoic acid, 99%

Catalog No. AC109681000
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109681000 100 g Plastic Bottle
AC109680100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109681000 Supplier Thermo Scientific Chemicals Supplier No. 109681000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2516-95-2 | C7H4ClNO4 | 201.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2516-95-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.57
MDL Number MFCD00007290
InChI Key ZKUYSJHXBFFGPU-UHFFFAOYSA-N
Synonym 5-chloro-2-nitrobenzoic acid, benzoic acid, 5-chloro-2-nitro, 2-nitro-5-chlorobenzoic acid, 3-chloro-6-nitrobenzoic acid, 5-chloro-2-nitro-benzoic acid, pubchem4586, acmc-209ghs, rarechem al bo 1106, timtec-bb sbb009922, 2-nitro-5-chlorobenzoate
PubChem CID 17286
SMILES C1=CC(=C(C=C1Cl)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 136°C to 139°C
Color Green-Yellow to Yellow
Flash Point >100°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic Bottle
Linear Formula ClC6H3(NO2)CO2H
Quantity 100 g
Beilstein 09, 401
Solubility Information Solubility in water: 9,67g/L (25°C). Other solubilities: soluble in methanol
Formula Weight 201.57
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitrobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 219-738-1

RUO – Research Use Only