missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitrobenzoic acid, 99%

Catalog No. AAA1249306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1249306 5 g
AAA1249314 25 g
AAA1249322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1249306 Supplier Thermo Scientific Chemicals Supplier No. A1249306

CAS: 2516-95-2 | C7H4ClNO4 | 201.562 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2516-95-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007290
InChI Key ZKUYSJHXBFFGPU-UHFFFAOYSA-N
Synonym 5-chloro-2-nitrobenzoic acid, benzoic acid, 5-chloro-2-nitro, 2-nitro-5-chlorobenzoic acid, 3-chloro-6-nitrobenzoic acid, 5-chloro-2-nitro-benzoic acid, pubchem4586, acmc-209ghs, rarechem al bo 1106, timtec-bb sbb009922, 2-nitro-5-chlorobenzoate
PubChem CID 17286
SMILES C1=CC(=C(C=C1Cl)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 136°C to 140°C
Odor Odorless
Quantity 5 g
Beilstein 1963952
Formula Weight 201.57
Percent Purity 99%
Chemical Name or Material 5-Chloro-2-nitrobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 219-738-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only