missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitroaniline 99.0+%, TCI America™

Catalog No. p-7124813
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C113925G 25 g
C1139250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C113925G Supplier TCI America Supplier No. C113925G
Only null left

Chemical Identifiers

CAS 1635-61-6
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007776
InChI Key ZCWXYZBQDNFULS-UHFFFAOYSA-N
Synonym 2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline
PubChem CID 74218
IUPAC Name 5-chloro-2-nitroaniline
SMILES C1=CC(=C(C=C1Cl)N)[N+](=O)[O-]

Specifications

Melting Point 128°C
Quantity 25 g
UN Number 2237
Formula Weight 172.57
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitroaniline
EINECSNumber (3)-0407
TSCA No