missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD04371DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CD04371DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

35375-74-7
240.566
AAMCWLIPRMIPBN-UHFFFAOYSA-N
2773813
C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
C7H4ClF3N2O2
MFCD01312277
5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803
5-chloro-2-nitro-4-(trifluoromethyl)aniline

Specifications

5-chloro-2-nitro-4-(trifluoromethyl)aniline
96.5
97%
C7H4ClF3N2O2
MFCD01312277
AAMCWLIPRMIPBN-UHFFFAOYSA-N
5-chloro-2-nitro-4-(trifluoromethyl)aniline
2773813
97%
35375-74-7
100.0
Amber Glass Bottle
1g
5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803
C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
240.566
240.57
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions