missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog No. p-2701306
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD04371CB 250mg Amber Glass Bottle
CD04371DA 1g Amber Glass Bottle
CD04371DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD04371CB Supplier Thermo Scientific Chemicals Supplier No. CD04371CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35375-74-7
Molecular Formula C7H4ClF3N2O2
Molecular Weight (g/mol) 240.566
MDL Number MFCD01312277
InChI Key AAMCWLIPRMIPBN-UHFFFAOYSA-N
Synonym 5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803
PubChem CID 2773813
IUPAC Name 5-chloro-2-nitro-4-(trifluoromethyl)aniline
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 240.57
Percent Purity 97%
Chemical Name or Material 5-chloro-2-nitro-4-(trifluoromethyl)aniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.