missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylphenol, 98%

CAS: 5306-98-9 | C7H7ClO | 142.58 g/mol

$137.86 - $460.74

Chemical Identifiers

CAS 5306-98-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00060672
InChI Key KKFPXGXMSBBNJI-UHFFFAOYSA-N
Synonym 5-chloro-o-cresol, 5-chloro-2-cresol, o-cresol, 5-chloro, phenol, 5-chloro-2-methyl, 3-chloro-6-methylphenol, 2-methyl-5-chlorophenol, 4-chloro-2-hydroxytoluene, 5-chloro-2-methyl-phenol, phenol,5-chloro-2-methyl
PubChem CID 79192
IUPAC Name 5-chloro-2-methylphenol
SMILES CC1=CC=C(Cl)C=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2552706
View Documents
Thermo Scientific Chemicals
B2552706
5 g
Each for $137.86
Only null left
 
AAB2552714
View Documents
Thermo Scientific Chemicals
B2552714
25 g
Each for $460.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5306-98-9
142.58
KKFPXGXMSBBNJI-UHFFFAOYSA-N
79192
CC1=CC=C(Cl)C=C1O
C7H7ClO
MFCD00060672
5-chloro-o-cresol, 5-chloro-2-cresol, o-cresol, 5-chloro, phenol, 5-chloro-2-methyl, 3-chloro-6-methylphenol, 2-methyl-5-chlorophenol, 4-chloro-2-hydroxytoluene, 5-chloro-2-methyl-phenol, phenol,5-chloro-2-methyl
5-chloro-2-methylphenol

Specifications

5306-98-9
109°C to 112°C (15 mmHg)
MFCD00060672
UN3437
5-chloro-o-cresol, 5-chloro-2-cresol, o-cresol, 5-chloro, phenol, 5-chloro-2-methyl, 3-chloro-6-methylphenol, 2-methyl-5-chlorophenol, 4-chloro-2-hydroxytoluene, 5-chloro-2-methyl-phenol, phenol,5-chloro-2-methyl
CC1=CC=C(Cl)C=C1O
142.58
142.59
5-Chloro-2-methylphenol
70°C to 75°C
C7H7ClO
5 g
1906683
KKFPXGXMSBBNJI-UHFFFAOYSA-N
5-chloro-2-methylphenol
79192
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID

EINECSNumber : 226-160-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only