missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-methylbenzoxazole 98.0+%, TCI America™
$91.33 - $91.33
Chemical Identifiers
| CAS | 19219-99-9 |
|---|---|
| Molecular Formula | C8H6ClNO |
| Molecular Weight (g/mol) | 167.592 |
| MDL Number | MFCD00022850 |
| InChI Key | HJCIGAUHTJBHBQ-UHFFFAOYSA-N |
| PubChem CID | 29510 |
| IUPAC Name | 5-chloro-2-methyl-1,3-benzoxazole |
| SMILES | CC1=NC2=C(O1)C=CC(=C2)Cl |
Chemical Identifiers
| 19219-99-9 | |
| 167.592 | |
| HJCIGAUHTJBHBQ-UHFFFAOYSA-N | |
| 5-chloro-2-methyl-1,3-benzoxazole |
| C8H6ClNO | |
| MFCD00022850 | |
| 29510 | |
| CC1=NC2=C(O1)C=CC(=C2)Cl |
Specifications
| 19219-99-9 | |
| 220°C | |
| MFCD00022850 | |
| HJCIGAUHTJBHBQ-UHFFFAOYSA-N | |
| 5-chloro-2-methyl-1,3-benzoxazole | |
| 29510 | |
| ≥98.0% (GC) | |
| 5-Chloro-2-methylbenzoxazole |
| 57°C | |
| C8H6ClNO | |
| 25 g | |
| CC1=NC2=C(O1)C=CC(=C2)Cl | |
| 167.592 | |
| 167.59 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DM4790000
TSCA : Yes