accessibility menu, dialog, popup

UserName

CAS RN 19219-99-9

IUPAC Name: 5-chloro-2-methyl-1,3-benzoxazole
Synonyms: 5-chloro-2-methyl-1,3-benzoxazole
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: HJCIGAUHTJBHBQ-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(Cl)=CC=C2O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results