missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylbenzoxazole 98.0+%, TCI America™

Catalog No. C127625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C127625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C127625G Supplier TCI America Supplier No. C127625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19219-99-9
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00022850
InChI Key HJCIGAUHTJBHBQ-UHFFFAOYSA-N
PubChem CID 29510
IUPAC Name 5-chloro-2-methyl-1,3-benzoxazole
SMILES CC1=NC2=C(O1)C=CC(=C2)Cl

Specifications

Melting Point 57°C
Boiling Point 220°C
Quantity 25 g
Formula Weight 167.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methylbenzoxazole
RTECSNumber DM4790000
TSCA Yes