missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylbenzothiazole 99.0+%, TCI America™

Catalog No. c147525g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C147525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C147525G Supplier TCI America Supplier No. C147525G

Chemical Identifiers

CAS 1006-99-1
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.653
MDL Number MFCD00022881
InChI Key XCALAYIRFYALSX-UHFFFAOYSA-N
Synonym 5-chloro-2-methylbenzothiazole, 5-chloro-2-methylbenzo d thiazole, 2-methyl-5-chlorobenzothiazole, benzothiazole, 5-chloro-2-methyl, usaf ek-p-4382, 5-chloro-2-methyl-benzothiazole, 2-methyl-5-chloro benzothiazole, 5-chloro-2-methyl benzothiazole, 2-metyl-5-chloro-benzothiazole, pubchem10878
PubChem CID 13873
IUPAC Name 5-chloro-2-methyl-1,3-benzothiazole
SMILES CC1=NC2=C(S1)C=CC(=C2)Cl

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 147°C
Quantity 25 g
Formula Weight 183.65
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methylbenzothiazole
EINECSNumber (5)-0230
RTECSNumber DL3325000
TSCA Yes