missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. C25095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25095G 5 g
C250925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25095G Supplier TCI America Supplier No. C25095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7499-06-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045842
InChI Key CSAPESWNZDOAFU-UHFFFAOYSA-N
Synonym 2-methyl-5-chlorobenzoic acid, 5-chloro-o-toluic acid, 5-chloro-2-methyl-benzoic acid, benzoic acid, 5-chloro-2-methyl, o-toluic acid, 5-chloro, 5-chloro-2-methylbenzoicacid, 5-chloro-2-methyl benzoic acid, 3-chloro-6-methylbenzoic acid, pubchem4578, ksc173q5r
PubChem CID 82009
IUPAC Name 5-chloro-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)Cl)C(=O)O

Specifications

Melting Point 173°C
Quantity 5 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methylbenzoic Acid