missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylaniline, 98%

Catalog No. AC109380050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109380050 5 g Glass bottle
AC109381000 100 g Glass bottle
AC109385000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC109380050 Supplier Thermo Scientific Chemicals Supplier No. 109380050

CAS: 95-79-4 | C7H8ClN | 141.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-79-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007779
InChI Key WRZOMWDJOLIVQP-UHFFFAOYSA-N
Synonym 5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb
PubChem CID 7260
ChEBI CHEBI:82422
IUPAC Name 5-chloro-2-methylaniline
SMILES CC1=CC=C(Cl)C=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 22°C
Color Yellow to Brown
Boiling Point 237°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5830 to 1.585
Linear Formula ClC6H3(CH3)NH2
Quantity 5 g
Beilstein 12, 835
Solubility Information Solubility in water: < 1g/L (22°C). Other solubilities: soluble in alcohol,ether,benzene,chloroform and,dioxane
Formula Weight 141.6
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 5-Chloro-2-methylaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 202-452-6
RTECSNumber XU5075000
TSCA TSCA
Recommended Storage Product may darken on storage

RUO – Research Use Only