missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylaniline 98.0+%, TCI America™

$105.89 - $105.89

Chemical Identifiers

CAS 95-79-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007779
InChI Key WRZOMWDJOLIVQP-UHFFFAOYSA-N
Synonym 5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb
PubChem CID 7260
ChEBI CHEBI:82422
IUPAC Name 5-chloro-2-methylaniline
SMILES CC1=CC=C(Cl)C=C1N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C187625G
View Documents
TCI America
C187625G
25 g
Each for $105.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

95-79-4
141.60
WRZOMWDJOLIVQP-UHFFFAOYSA-N
7260
5-chloro-2-methylaniline
C7H8ClN
MFCD00007779
5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb
CHEBI:82422
CC1=CC=C(Cl)C=C1N

Specifications

95-79-4
237°C
MFCD00007779
2239
WRZOMWDJOLIVQP-UHFFFAOYSA-N
5-chloro-2-methylaniline
7260
141.60
Crystalline Lumps
White-Yellow
C7H8ClN
25 g
5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb
CC1=CC=C(Cl)C=C1N
141.60
CHEBI:82422
≥98.0% (GC,T)
5-Chloro-2-methylaniline
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0265

RTECSNumber : XU5075000

TSCA : Yes

SDS
Documents

Documents