missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(methylamino)benzophenone 98.0+%, TCI America™

Catalog No. C228125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C228125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C228125G Supplier TCI America Supplier No. C228125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1022-13-5
Molecular Formula C14H12ClNO
Molecular Weight (g/mol) 245.71
MDL Number MFCD00008284
InChI Key WPNMLCMTDCANOZ-UHFFFAOYSA-N
Synonym 5-chloro-2-methylamino benzophenone, 5-chloro-2-methylaminobenzophenone, 2-methylamino-5-chlorobenzophenone, 5-chloro-2-methylamino phenyl phenyl methanone, methanone, 5-chloro-2-methylamino phenyl phenyl, unii-d4gd5770pi, benzophenone, 5-chloro-2-methylamino, 2-benzoyl-4-chloro-n-methylaniline, 5-chloro-2-methylamino phenyl phenylmethanone, 5-chloro-2-methylamino phenyl-phenylmethanone
PubChem CID 13925
IUPAC Name 2-benzoyl-4-chloro-N-methylaniline
SMILES CNC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1

Specifications

Melting Point 97°C
Color Yellow
Quantity 25 g
Formula Weight 245.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-(methylamino)benzophenone
RTECSNumber DJ0414000
TSCA Yes