missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxyaniline 98.0+%, TCI America™

Catalog No. C011825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C011825G 25 g
C0118500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C011825G Supplier TCI America Supplier No. C011825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-03-4
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00007777
InChI Key WBSMIPLNPSCJFS-UHFFFAOYSA-N
Synonym 2-amino-4-chloroanisole, 5-chloro-o-anisidine, benzenamine, 5-chloro-2-methoxy, 2-methoxy-5-chloroaniline, tulabase fast red r, tulabase fast red rc, 4-chloro-2-aminoanisole, o-anisidine, 5-chloro, 5-chloro-2-methoxyphenylamine, p-chloro-o-aminoanisole
PubChem CID 66763
IUPAC Name 5-chloro-2-methoxyaniline
SMILES COC1=C(C=C(C=C1)Cl)N

Specifications

Melting Point 82°C
Quantity 25 g
UN Number 2233
Formula Weight 157.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methoxyaniline
EINECSNumber (3)-0908&(3)-0971
TSCA Yes