missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxyaniline, 98%

Catalog No. p-7021772
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1424422 100 g
AAA1424436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1424422 Supplier Thermo Scientific Chemicals Supplier No. A1424422

CAS: 95-03-4 | C7H8ClNO | 157.597 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-03-4
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00007777
InChI Key WBSMIPLNPSCJFS-UHFFFAOYSA-N
Synonym 2-amino-4-chloroanisole, 5-chloro-o-anisidine, benzenamine, 5-chloro-2-methoxy, 2-methoxy-5-chloroaniline, tulabase fast red r, tulabase fast red rc, 4-chloro-2-aminoanisole, o-anisidine, 5-chloro, 5-chloro-2-methoxyphenylamine, p-chloro-o-aminoanisole
PubChem CID 66763
IUPAC Name 5-chloro-2-methoxyaniline
SMILES COC1=C(C=C(C=C1)Cl)N

Specifications

Melting Point 79°C to 84°C
Boiling Point 135°C (8 mmHg)
Odor Characteristic
Quantity 100 g
UN Number UN2233
Beilstein 879161
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methoxyaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROANISIDINES
EINECSNumber 202-385-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only