missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-mercaptobenzothiazole, 98%

Catalog No. p-7023323
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1674706 5 g
AAA1674714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1674706 Supplier Thermo Scientific Chemicals Supplier No. A1674706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5331-91-9 | C7H4ClNS2 | 201.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5331-91-9
Molecular Formula C7H4ClNS2
Molecular Weight (g/mol) 201.69
MDL Number MFCD00005783
InChI Key NKYDKCVZNMNZCM-UHFFFAOYSA-N
Synonym 5-chloro-2-mercaptobenzothiazole, 5-chloro-1,3-benzothiazole-2-thiol, 5-chlorobenzo d thiazole-2 3h-thione, sh-benzothiazole, 5-chloro-2-benzothiazolethiol, 2 3h-benzothiazolethione, 5-chloro, 5-chlorobenzo d thiazole-2-thiol, 5-chlorobenzothiazole-2-thiol, benzothiazole, 5-chloro-2-mercapto, 2-benzothiazolethiol, 5-chloro
PubChem CID 2723842
IUPAC Name 5-chloro-3H-1,3-benzothiazole-2-thione
SMILES ClC1=CC=C2SC(=S)NC2=C1

Specifications

Melting Point 198°C to 200°C
Quantity 5 g
Formula Weight 201.7
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-mercaptobenzothiazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-235-0
RTECSNumber DL6490000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only