accessibility menu, dialog, popup

UserName

ClSSHN

CAS RN 5331-91-9

IUPAC Name: 5-chloro-2,3-dihydro-1,3-benzothiazole-2-thione
Synonyms: 5-chloro-3H-1,3-benzothiazole-2-thione
Molecular Weight (g/mol): 201.69
Molecular Formula: C7H4ClNS2
InChi Key: NKYDKCVZNMNZCM-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SC(=S)NC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)