missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-mercaptobenzimidazole 98.0+%, TCI America™
Supplier: TCI America C159025G
Specifications
| 5-Chloro-2-mercaptobenzimidazole | |
| 294°C | |
| MFCD01658762 | |
| 2811 | |
| ZZIHEYOZBRPWMB-UHFFFAOYSA-N | |
| 5-chloro-1,3-dihydrobenzimidazole-2-thione | |
| 2056429 | |
| ≥98.0% (T) |
| 25369-78-2 | |
| C7H5ClN2S | |
| 25 g | |
| 5-chloro-2-mercaptobenzimidazole, 6-chloro-1h-benzimidazole-2-thiol, 5-chloro-2-benzimidazolethiol, 2-benzimidazolinethione, 5-chloro, 5-chloro-1,3-dihydro-2h-benzimidazole-2-thione, 5-chloro-1h-benzimidazole-2-thiol, 2-benzimidazolethiol, 5-chloro, 5-chloro-2 3-benzimidazolethione, 6-chloro-1h-benzoimidazole-2-thiol, 6-chloro-1h-benzo d imidazole-2-thiol | |
| C1=CC2=C(C=C1Cl)NC(=S)N2 | |
| 184.641 | |
| 184.64 | |
| Crystalline Powder |
Chemical Identifiers
| 25369-78-2 | |
| 184.641 | |
| ZZIHEYOZBRPWMB-UHFFFAOYSA-N | |
| 2056429 | |
| C1=CC2=C(C=C1Cl)NC(=S)N2 |
| C7H5ClN2S | |
| MFCD01658762 | |
| 5-chloro-2-mercaptobenzimidazole, 6-chloro-1h-benzimidazole-2-thiol, 5-chloro-2-benzimidazolethiol, 2-benzimidazolinethione, 5-chloro, 5-chloro-1,3-dihydro-2h-benzimidazole-2-thione, 5-chloro-1h-benzimidazole-2-thiol, 2-benzimidazolethiol, 5-chloro, 5-chloro-2 3-benzimidazolethione, 6-chloro-1h-benzoimidazole-2-thiol, 6-chloro-1h-benzo d imidazole-2-thiol | |
| 5-chloro-1,3-dihydrobenzimidazole-2-thione |
Safety and Handling
RTECSNumber : DE1605000
TSCA : No