missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-mercaptobenzimidazole 98.0+%, TCI America™

Catalog No. p-7125076
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C15905G 5 g
C159025G 25 g
2 options

Catalog No. C15905G

Supplier: TCI America C15905G

Only null left

Chemical Identifiers

CAS 25369-78-2
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.641
MDL Number MFCD01658762
InChI Key ZZIHEYOZBRPWMB-UHFFFAOYSA-N
Synonym 5-chloro-2-mercaptobenzimidazole, 6-chloro-1h-benzimidazole-2-thiol, 5-chloro-2-benzimidazolethiol, 2-benzimidazolinethione, 5-chloro, 5-chloro-1,3-dihydro-2h-benzimidazole-2-thione, 5-chloro-1h-benzimidazole-2-thiol, 2-benzimidazolethiol, 5-chloro, 5-chloro-2 3-benzimidazolethione, 6-chloro-1h-benzoimidazole-2-thiol, 6-chloro-1h-benzo d imidazole-2-thiol
PubChem CID 2056429
IUPAC Name 5-chloro-1,3-dihydrobenzimidazole-2-thione
SMILES C1=CC2=C(C=C1Cl)NC(=S)N2

Specifications

Melting Point 294°C
Quantity 5 g
UN Number 2811
Formula Weight 184.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-mercaptobenzimidazole
RTECSNumber DE1605000
TSCA No