Learn More
5-Chloro-2-mercaptobenzimidazole, 98%
CAS: 25369-78-2 | C7H5ClN2S | 184.641 g/mol
Supplier: Thermo Scientific Chemicals A1985214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Chloro-2-mercaptobenzimidazole | |
| 298°C to 302°C | |
| MFCD01658762 | |
| UN2811 | |
| ZZIHEYOZBRPWMB-UHFFFAOYSA-N | |
| 5-chloro-1,3-dihydrobenzimidazole-2-thione | |
| 2056429 | |
| 98% |
| 25369-78-2 | |
| C7H5ClN2S | |
| 25 g | |
| 5-chloro-2-mercaptobenzimidazole, 6-chloro-1h-benzimidazole-2-thiol, 5-chloro-2-benzimidazolethiol, 2-benzimidazolinethione, 5-chloro, 5-chloro-1,3-dihydro-2h-benzimidazole-2-thione, 5-chloro-1h-benzimidazole-2-thiol, 2-benzimidazolethiol, 5-chloro, 5-chloro-2 3-benzimidazolethione, 6-chloro-1h-benzoimidazole-2-thiol, 6-chloro-1h-benzo d imidazole-2-thiol | |
| C1=CC2=C(C=C1Cl)NC(=S)N2 | |
| 184.641 | |
| 184.64 |
Chemical Identifiers
| 25369-78-2 | |
| 184.641 | |
| ZZIHEYOZBRPWMB-UHFFFAOYSA-N | |
| 2056429 | |
| C1=CC2=C(C=C1Cl)NC(=S)N2 |
| C7H5ClN2S | |
| MFCD01658762 | |
| 5-chloro-2-mercaptobenzimidazole, 6-chloro-1h-benzimidazole-2-thiol, 5-chloro-2-benzimidazolethiol, 2-benzimidazolinethione, 5-chloro, 5-chloro-1,3-dihydro-2h-benzimidazole-2-thione, 5-chloro-1h-benzimidazole-2-thiol, 2-benzimidazolethiol, 5-chloro, 5-chloro-2 3-benzimidazolethione, 6-chloro-1h-benzoimidazole-2-thiol, 6-chloro-1h-benzo d imidazole-2-thiol | |
| 5-chloro-1,3-dihydrobenzimidazole-2-thione |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 246-903-5
RTECSNumber : DE1605000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only