missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-mercaptobenzimidazole, 98%

Catalog No. p-7024700
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1985206 5 g
AAA1985214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1985206 Supplier Thermo Scientific Chemicals Supplier No. A1985206
Only null left

CAS: 25369-78-2 | C7H5ClN2S | 184.641 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25369-78-2
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.641
MDL Number MFCD01658762
InChI Key ZZIHEYOZBRPWMB-UHFFFAOYSA-N
Synonym 5-chloro-2-mercaptobenzimidazole, 6-chloro-1h-benzimidazole-2-thiol, 5-chloro-2-benzimidazolethiol, 2-benzimidazolinethione, 5-chloro, 5-chloro-1,3-dihydro-2h-benzimidazole-2-thione, 5-chloro-1h-benzimidazole-2-thiol, 2-benzimidazolethiol, 5-chloro, 5-chloro-2 3-benzimidazolethione, 6-chloro-1h-benzoimidazole-2-thiol, 6-chloro-1h-benzo d imidazole-2-thiol
PubChem CID 2056429
IUPAC Name 5-chloro-1,3-dihydrobenzimidazole-2-thione
SMILES C1=CC2=C(C=C1Cl)NC(=S)N2

Specifications

Melting Point 298°C to 302°C
Quantity 5 g
UN Number UN2811
Formula Weight 184.64
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-mercaptobenzimidazole
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 246-903-5
RTECSNumber DE1605000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only