missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxybenzyl alcohol, 98%

Catalog No. p-7031451
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3253303 1 g
AAH3253309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3253303 Supplier Thermo Scientific Chemicals Supplier No. H3253303
Only null left

CAS: 5330-38-1 | C7H7ClO2 | 158.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5330-38-1
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
MDL Number MFCD00021967
InChI Key AFPSETMCMFRVPG-UHFFFAOYSA-N
Synonym 5-chlorosalicyl alcohol, 4-chloro-2-hydroxymethyl phenol, 5-chloro-2-hydroxybenzyl alcohol, benzenemethanol, 5-chloro-2-hydroxy, benzenemethanol,5-chloro-2-hydroxy, 5-chlorosaligenin, 5-chlorosalicylalcohol, saligenin, 5-chloro, 5-chloro-2-hydroxyphenyl methanol, 5-chloro-2-hydroxyphenylmethyl carbinol
PubChem CID 79238
IUPAC Name 4-chloro-2-(hydroxymethyl)phenol
SMILES C1=CC(=C(C=C1Cl)CO)O

Specifications

Melting Point 84°C to 87°C
Quantity 1 g
Formula Weight 158.58
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-hydroxybenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only