missing translation for 'onlineSavingsMsg'
Learn More

5'-Chloro-2'-hydroxy-4'-methylacetophenone 95.0+%, TCI America™

Catalog No. C16295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16295G 5 g
C162925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16295G Supplier TCI America Supplier No. C16295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28480-70-8
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00191920
InChI Key HDUSGGZSLVCDKY-UHFFFAOYSA-N
Synonym 4-Acetyl-2-chloro-5-hydroxytoluene, 2-Acetyl-4-chloro-5-methylphenol
PubChem CID 598229
IUPAC Name 1-(5-chloro-2-hydroxy-4-methylphenyl)ethanone
SMILES CC1=C(C=C(C(=C1)O)C(=O)C)Cl

Specifications

Melting Point 69°C
Color Yellow
Boiling Point 137°C
Quantity 5 g
Formula Weight 184.62
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Chloro-2′-hydroxy-4′-methylacetophenone