accessibility menu, dialog, popup

UserName

CH3HOOH3CCl

CAS RN 28480-70-8

IUPAC Name: 1-(5-chloro-2-hydroxy-4-methylphenyl)ethan-1-one
Synonyms: 1-(5-chloro-2-hydroxy-4-methylphenyl)ethanone
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: HDUSGGZSLVCDKY-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=C(C)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results