missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxy-3-methylbenzaldehyde, 95%, Thermo Scientific™

Chemical Identifiers

CAS 23602-63-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00275687
InChI Key NSKZAOKQZDLHGO-UHFFFAOYSA-N
Synonym benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
PubChem CID 2774358
IUPAC Name 5-chloro-2-hydroxy-3-methylbenzaldehyde
SMILES CC1=C(C(=CC(=C1)Cl)C=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB03914CB
SDS View Documents
Thermo Scientific Chemicals
BTB03914CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB03914DA
SDS View Documents
Thermo Scientific Chemicals
BTB03914DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

23602-63-3
170.592
NSKZAOKQZDLHGO-UHFFFAOYSA-N
2774358
CC1=C(C(=CC(=C1)Cl)C=O)O
C8H7ClO2
MFCD00275687
benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
5-chloro-2-hydroxy-3-methylbenzaldehyde

Specifications

23602-63-3
100.0
Amber Glass Bottle
MFCD00275687
benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
CC1=C(C(=CC(=C1)Cl)C=O)O
170.592
170.59
5-Chloro-2-hydroxy-3-methylbenzaldehyde
94.5
95%
C8H7ClO2
250mg
NSKZAOKQZDLHGO-UHFFFAOYSA-N
5-chloro-2-hydroxy-3-methylbenzaldehyde
2774358
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'