missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-furaldehyde, 98%, stab. with 2% ethanol

Catalog No. AAL1997103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1997103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1997103 Supplier Thermo Scientific Chemicals Supplier No. L1997103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21508-19-0 | C5H3ClO2 | 130.527 g/mol

(+)-1-Deoxynojirimycin hydrochloride interferes with normal processing of N-linked glycoproteins, producing reduced amounts of complex type oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-1-Deoxynojirimycin hydrochloride interferes with normal processing of N-linked glycoproteins, producing reduced amounts of complex type oligosaccharides.

Solubility
Insoluble in water.

Notes
Air

Chemical Identifiers

CAS 21508-19-0
Molecular Formula C5H3ClO2
Molecular Weight (g/mol) 130.527
MDL Number MFCD02752599
InChI Key DGAUAVDWXYXXGQ-UHFFFAOYSA-N
Synonym 5-chloro-2-furaldehyde, 5-chlorofurfural, 2-furancarboxaldehyde, 5-chloro, 5-chloro-furfural, 2-chlorofuran-5-aldehyde, acmc-1cq7z, 5-chloro-furan-2carbaldehyde, 5-chlorofuran-2 carbaldehyde, 5-chloro-furan-2-carbaldehyde, 5-chlorofuran-2-carboxaldehyde
PubChem CID 2769630
IUPAC Name 5-chlorofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)Cl)C=O

Specifications

Melting Point 33°C to 36°C
Boiling Point 70°C (10 mmHg)
Quantity 1 g
Beilstein 108401
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 130.53
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-furaldehyde, stabilized with 2% ethanol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only