accessibility menu, dialog, popup

UserName

OClO

CAS RN 21508-19-0

IUPAC Name: 5-chlorofuran-2-carbaldehyde
Synonyms: 5-chlorofuran-2-carbaldehyde
Molecular Weight (g/mol): 130.53
Molecular Formula: C5H3ClO2
InChi Key: DGAUAVDWXYXXGQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(O1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results