missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C26455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26455G Supplier TCI America Supplier No. C26455G

Chemical Identifiers

CAS 352535-83-2
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD05664225
InChI Key GGTUVWGMCFXUAS-UHFFFAOYSA-N
Synonym 5-chloro-2-fluorophenyl boronic acid, 5-chloro-2-fluorobenzeneboronic acid, 5-chloro-2-fluorophenylboronicacid, 5-chloro-2-fluoro-phenyl boronic acid, boronic acid, 5-chloro-2-fluorophenyl, pubchem11819, acmc-1ae0a, boronic acid,b-5-chloro-2-fluorophenyl, boronic acid, b-5-chloro-2-fluorophenyl
PubChem CID 3718925
IUPAC Name (5-chloro-2-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(Cl)=CC=C1F

Specifications

Color White
Quantity 5 g
Formula Weight 174.36
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-fluorophenylboronic Acid (contains varying amounts of Anhydride)