missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluoronitrobenzene 98.0+%, TCI America™

Catalog No. C27045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27045G 5 g
C270425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27045G Supplier TCI America Supplier No. C27045G
Only null left

Chemical Identifiers

CAS 345-18-6
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.543
MDL Number MFCD00069418
InChI Key DIAWBHLTWNWYGR-UHFFFAOYSA-N
Synonym 4-Chloro-1-fluoro-2-nitrobenzene
PubChem CID 67660
IUPAC Name 4-chloro-1-fluoro-2-nitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])F

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 237°C
Quantity 5 g
Formula Weight 175.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-fluoronitrobenzene