missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-fluorobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America C31145G
Specifications
| 5-Chloro-2-fluorobenzaldehyde | |
| 37°C | |
| C7H4ClFO | |
| 5 g | |
| WDTUCEMLUHTMCB-UHFFFAOYSA-N | |
| 5-chloro-2-fluorobenzaldehyde | |
| 2773586 | |
| ≥98.0% (GC) |
| 96515-79-6 | |
| White-Yellow | |
| MFCD03094518 | |
| 2-fluoro-5-chlorobenzaldehyde, 3-chloro-6-fluoro benzaldehyde, 3-chloro-6-fluorobenzaldehyde, 5-chloro-2-fluoro-benzaldehyde, benzaldehyde, 5-chloro-2-fluoro, 2-fluoro-5-chloro-benzaldehyde, zlchem 333, pubchem1411, ksc486m0t, 2-fluoro-5-chloro benzaldehyde | |
| FC1=CC=C(Cl)C=C1C=O | |
| 158.56 | |
| 158.56 | |
| Crystalline Powder |
Chemical Identifiers
| 96515-79-6 | |
| 158.56 | |
| WDTUCEMLUHTMCB-UHFFFAOYSA-N | |
| 2773586 | |
| FC1=CC=C(Cl)C=C1C=O |
| C7H4ClFO | |
| MFCD03094518 | |
| 2-fluoro-5-chlorobenzaldehyde, 3-chloro-6-fluoro benzaldehyde, 3-chloro-6-fluorobenzaldehyde, 5-chloro-2-fluoro-benzaldehyde, benzaldehyde, 5-chloro-2-fluoro, 2-fluoro-5-chloro-benzaldehyde, zlchem 333, pubchem1411, ksc486m0t, 2-fluoro-5-chloro benzaldehyde | |
| 5-chloro-2-fluorobenzaldehyde |
Safety and Handling
TSCA : No