missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzaldehyde, 97%

Catalog No. AAH2618703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2618703 1 g
AAH2618706 5 g
AAH2618714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2618703 Supplier Thermo Scientific Chemicals Supplier No. H2618703

CAS: 96515-79-6 | C7H4ClFO | 158.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96515-79-6
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD03094518
InChI Key WDTUCEMLUHTMCB-UHFFFAOYSA-N
Synonym 2-fluoro-5-chlorobenzaldehyde, 3-chloro-6-fluoro benzaldehyde, 3-chloro-6-fluorobenzaldehyde, 5-chloro-2-fluoro-benzaldehyde, benzaldehyde, 5-chloro-2-fluoro, 2-fluoro-5-chloro-benzaldehyde, zlchem 333, pubchem1411, ksc486m0t, 2-fluoro-5-chloro benzaldehyde
PubChem CID 2773586
IUPAC Name 5-chloro-2-fluorobenzaldehyde
SMILES FC1=CC=C(Cl)C=C1C=O

Specifications

Melting Point 29°C to 32°C
Refractive Index 1.541
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 158.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only