missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-fluoroaniline 97.0+%, TCI America™
$78.93 - $234.87
Chemical Identifiers
| CAS | 2106-05-0 |
|---|---|
| Molecular Formula | C6H5ClFN |
| Molecular Weight (g/mol) | 145.561 |
| MDL Number | MFCD00069416 |
| InChI Key | JCYROOANFKVAIB-UHFFFAOYSA-N |
| Synonym | 2-fluoro-5-chloroaniline, 5-chloro-2-fluorophenylamine, benzenamine, 5-chloro-2-fluoro, buttpark 44\01-98, 5-chlor-2-fluoranilin, pubchem1515, 5-chloro-2-fluoroani, acmc-209fh5, 5-chloro-2-fluoro-aniline, 5-chloro-2-fluorobenzenamine |
| PubChem CID | 75015 |
| IUPAC Name | 5-chloro-2-fluoroaniline |
| SMILES | C1=CC(=C(C=C1Cl)N)F |
Chemical Identifiers
| 2106-05-0 | |
| 145.561 | |
| JCYROOANFKVAIB-UHFFFAOYSA-N | |
| 75015 | |
| C1=CC(=C(C=C1Cl)N)F |
| C6H5ClFN | |
| MFCD00069416 | |
| 2-fluoro-5-chloroaniline, 5-chloro-2-fluorophenylamine, benzenamine, 5-chloro-2-fluoro, buttpark 44\01-98, 5-chlor-2-fluoranilin, pubchem1515, 5-chloro-2-fluoroani, acmc-209fh5, 5-chloro-2-fluoro-aniline, 5-chloro-2-fluorobenzenamine | |
| 5-chloro-2-fluoroaniline |
Specifications
| 2106-05-0 | |
| 105°C | |
| Clear Liquid at 20°C | |
| MFCD00069416 | |
| JCYROOANFKVAIB-UHFFFAOYSA-N | |
| 5-chloro-2-fluoroaniline | |
| 75015 | |
| ≥97.0% (GC) |
| Yellow | |
| 5 g | |
| C6H5ClFN | |
| 2-fluoro-5-chloroaniline, 5-chloro-2-fluorophenylamine, benzenamine, 5-chloro-2-fluoro, buttpark 44\01-98, 5-chlor-2-fluoranilin, pubchem1515, 5-chloro-2-fluoroani, acmc-209fh5, 5-chloro-2-fluoro-aniline, 5-chloro-2-fluorobenzenamine | |
| C1=CC(=C(C=C1Cl)N)F | |
| 145.561 | |
| 145.56 | |
| 5-Chloro-2-fluoroaniline |
Safety and Handling
TSCA : No