missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-adamantanone 98.0+%, TCI America™

Catalog No. C21401G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C21401G Supplier TCI America Supplier No. C21401G
Only null left

Chemical Identifiers

CAS 20098-17-3
Molecular Formula C10H13ClO
Molecular Weight (g/mol) 184.663
MDL Number MFCD00798599
InChI Key JPEOUSFBWXVGFX-UHFFFAOYSA-N
Synonym 5-chloro-2-adamantanone, 5-chloro-2-adamantone, adamantan-4-one, 1-chloro, acmc-1cljc, 5-chloro-2 adamantanone, 8-chloroadamantan-2-one, 5-chloroadamantane-2-one, 5-chloranyladamantan-2-one, tricyclo 3.3.1.1∼3,7∼ decan-2-one, 5-chloro
PubChem CID 334400
IUPAC Name 5-chloroadamantan-2-one
SMILES C1C2CC3CC(C2)(CC1C3=O)Cl

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 1 g
Formula Weight 184.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-adamantanone