CAS RN 20098-17-3
IUPAC Name:
(1R,3S)-5-chloroadamantan-2-one
Synonyms:
(1R,3S)-5-chloroadamantan-2-one
Molecular Weight (g/mol):
184.66
Molecular Formula:
C10H13ClO
InChi Key:
JPEOUSFBWXVGFX-VZCHMASFSA-N
SMILES:
ClC12CC3C[C@@H](C1)C(=O)[C@H](C3)C2
1
–
1
of
1
results
1
5-Chloro-2-adamantanone 98.0+%, TCI America™
CAS: 20098-17-3 Molecular Formula: C10H13ClO Molecular Weight (g/mol): 184.663 MDL Number: MFCD00798599 InChI Key: JPEOUSFBWXVGFX-UHFFFAOYSA-N Synonym: 5-chloro-2-adamantanone,5-chloro-2-adamantone,adamantan-4-one, 1-chloro,acmc-1cljc,5-chloro-2 adamantanone,8-chloroadamantan-2-one,5-chloroadamantane-2-one,5-chloranyladamantan-2-one,tricyclo 3.3.1.1∼3,7∼ decan-2-one, 5-chloro PubChem CID: 334400 IUPAC Name: 5-chloroadamantan-2-one SMILES: C1C2CC3CC(C2)(CC1C3=O)Cl