missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2,4-dinitrotoluene, 97%

Catalog No. AAL1180806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1180806 5 g
AAL1180814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1180806 Supplier Thermo Scientific Chemicals Supplier No. L1180806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51676-74-5 | C7H5ClN2O4 | 216.577 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51676-74-5
Molecular Formula C7H5ClN2O4
Molecular Weight (g/mol) 216.577
MDL Number MFCD00041235
InChI Key KPDPGZNHKMJEFZ-UHFFFAOYSA-N
Synonym 5-chloro-2,4-dinitrotoluene, toluene, 5-chloro-2,4-dinitro, 1-chloro-5-methyl-2,4-dinitro-benzene, 2-chloro-4-methyl-1,5-dinitrobenzene, acmc-20am4s, 5-chlor-2.4-dinitrotoluol, benzene,1-chloro-5-methyl-2,4-dinitro, 1-chloranyl-5-methyl-2,4-dinitro-benzene
PubChem CID 253802
IUPAC Name 1-chloro-5-methyl-2,4-dinitrobenzene
SMILES CC1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Cl

Specifications

Melting Point 89°C to 91°C
Quantity 5 g
UN Number UN2811
Beilstein 2506591
Formula Weight 216.58
Percent Purity 97%
Chemical Name or Material 5-Chloro-2,4-dinitrotoluene
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only