missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 97.0+%, TCI America™

Catalog No. p-7125918
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C33011G 1 g
C33015G 5 g
2 options

Catalog No. C33011G

Supplier: TCI America C33011G

Only null left

Chemical Identifiers

CAS 635305-24-7
Molecular Formula C10H14BClO2S
Molecular Weight (g/mol) 244.54
MDL Number MFCD08063140
InChI Key DDDRRTOIHWNUSI-UHFFFAOYSA-N
Synonym 2-(5-Chloro-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-Chloro-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 10354494
IUPAC Name 2-(5-chlorothiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)Cl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 244.54
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene